2-Fluor-6-methoxyphenylboronsäure 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-6-methoxybenzeneboronic acid
PC1639-100G
PC1639-25G
PC1639-5G
APOSPC1639-100GEA
227
EUR
APOSPC1639-100G
APOSPC1639-25G
APOSPC1639-5G
2-Fluor-6-methoxyphenylboronsäure 99%
2-Fluor-6-methoxyphenylboronsäure
Formel:
C₇H₈BFO₃ Molecular Weight: 169,95 g/mol Schmelzpunkt: 124…126 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02179483 CAS-Nummer: 78495-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...