4-Carboxy-3-fluorphenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Borono-2-fluorobenzoic acid, 4-Carboxy-3-fluorobenzeneboronic acid
PC1644-25G
PC1644-5G
PC1644-100G
APOSPC1644-25GEA
186
EUR
APOSPC1644-25G
APOSPC1644-5G
APOSPC1644-100G
4-Carboxy-3-fluorphenylboronsäure 97%
4-Carboxy-3-fluorphenylboronsäure
Formel:
C₇H₆BFO₄ Molecular Weight: 183,93 g/mol Schmelzpunkt: 236…240 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01114671 CAS-Nummer: 120153-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...