2-Fluoro-5-trifluoromethylbenzyl chloride

Lieferant: Apollo Scientific
Gefahr

PC1670-25G PC1670-5G
APOSPC1670-25GEA 63.8 EUR
APOSPC1670-25G APOSPC1670-5G
2-Fluoro-5-trifluoromethylbenzyl chloride
2-Fluoro-5-trifluoromethylbenzyl chloride
Formel: C₈H₅ClF₄
Molecular Weight: 212.58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05864315
CAS-Nummer: 883543-26-8
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR