1-Boc-5-fluor-2-indolboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Boc-5-fluoroindole-2-boronic Acid, N-(Boc)-5-fluoroindole-2-boronic acid
PC1682-1G
PC1682-5G
APOSPC1682-1GEA
134
EUR
APOSPC1682-1G
APOSPC1682-5G
1-Boc-5-fluor-2-indolboronsäure 98%
1-Boc-5-fluor-2-indolboronsäure
Formel:
C₁₃H₁₅BFNO₄ Molecular Weight: 279.07 g/mol Schmelzpunkt: 130…140 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04114581 CAS-Nummer: 352359-23-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...