3-Brom-2-fluorphenol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Brom-2-fluor-3-hydroxybenzol
PC1687-1G
PC1687-25G
PC1687-5G
APOSPC1687-1GEA
77
EUR
APOSPC1687-1G
APOSPC1687-25G
APOSPC1687-5G
3-Brom-2-fluorphenol 97%
3-Brom-2-fluorphenol
Formel:
C₆H₄BrFO Schmelzpunkt: 52…54 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09864673 CAS-Nummer: 156682-53-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...