3',5'-Bis(trifluormethyl)-2-chloracetanilid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(Chloroacetyl)-3,5-bis(trifluoromethyl)aniline, N-(3,5-Bis(trifluoromethyl)phenyl)-2-chloroacetamide
PC1687H-1G
PC1687H-5G
PC1687H-250MG
APOSPC1687H-1GEA
29
EUR
APOSPC1687H-1G
APOSPC1687H-5G
APOSPC1687H-250MG
3',5'-Bis(trifluormethyl)-2-chloracetanilid 98%
3',5'-Bis(trifluormethyl)-2-chloracetanilid
Formel:
C₁₀H₆ClF₆NO Molecular Weight: 305.6042 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077474 CAS-Nummer: 790-75-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...