Kalium(4-(aminocarbonyl)phenyl)trifluoroborat 94%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Potassium 4-carbamoylphenyltrifluoroborate, Potassium (4-carbamoylphenyl)trifluoroborate
PC1703-1G
PC1703-5G
APOSPC1703-1GEA
34.8
EUR
APOSPC1703-1G
APOSPC1703-5G
Kalium(4-(aminocarbonyl)phenyl)trifluoroborat 94%
Kalium(4-(aminocarbonyl)phenyl)trifluoroborat
Formel:
C₇H₆BF₃KNO Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115758 CAS-Nummer: 850623-63-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...