Kalium(2-(benzyloxy)phenyl)trifluoroborat 95%
Lieferant: Apollo Scientific
PC1711-10G
PC1711-1G
PC1711-5G
APOSPC1711-10GEA
698
EUR
APOSPC1711-10G
APOSPC1711-1G
APOSPC1711-5G
Kalium(2-(benzyloxy)phenyl)trifluoroborat 95%
Kalium(2-(benzyloxy)phenyl)trifluoroborat
Formel:
C₁₃H₁₁BF₃KO Molecular Weight: 290.13 g/mol Schmelzpunkt: 200…204 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115742 CAS-Nummer: 850623-44-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...