Kalium(4-(benzyloxy)-2-methylphenyl)trifluoroborat 98%
Lieferant: Apollo Scientific
PC1721-1G
PC1721-5G
APOSPC1721-1GEA
152
EUR
APOSPC1721-1G
APOSPC1721-5G
Kalium(4-(benzyloxy)-2-methylphenyl)trifluoroborat 98%
Kalium(4-(benzyloxy)-2-methylphenyl)trifluoroborat
Formel:
C₁₄H₁₃BF₃KO Schmelzpunkt: 196…200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115740 CAS-Nummer: 850623-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...