Kalium(2-(hydroxymethyl)phenyl)trifluoroborat 96%

Lieferant: Apollo Scientific

PC1723-100G PC1723-25G PC1723-5G
APOSPC1723-100GEA 1490 EUR
APOSPC1723-100G APOSPC1723-25G APOSPC1723-5G
Kalium(2-(hydroxymethyl)phenyl)trifluoroborat 96%
Potassium-2-(hydroxymethyl)phenyltrifluoroborate
Formel: C₇H₇BF₃KO
Schmelzpunkt: 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115769
CAS-Nummer: 850623-74-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR