Kalium(3,4-dichlorphenyl)trifluoroborat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3,4-Dichlorphenyl)trifluorborsäure Kaliumsalz

PC1726-5G
APOSPC1726-5GEA 89.9 EUR
APOSPC1726-5G
Kalium(3,4-dichlorphenyl)trifluoroborat 98%
Kalium(3,4-dichlorphenyl)trifluoroborat
Formel: C₆H₃BCl₂F₃K
Molecular Weight: 252,9 g/mol
Schmelzpunkt: 178…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115763
CAS-Nummer: 850623-68-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR