Kalium(4-methoxy-2-methylphenyl)trifluoroborat 96%

Lieferant: Apollo Scientific

PC1729-1G PC1729-5G
APOSPC1729-1GEA 94.5 EUR
APOSPC1729-1G APOSPC1729-5G
Kalium(4-methoxy-2-methylphenyl)trifluoroborat 96%
Potassium-4-methoxy-2-methylphenyltrifluoroborate
Formel: C₈H₉BF₃KO
Lagertemperatur: Kühlschrank
MDL: MFCD04115764
CAS-Nummer: 850623-69-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR