5-Fluoroxindol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoroindolin-2-one, 5-Fluoro-2-oxoindoline, 5-Fluoro-2-oxindole

PC1735-5G PC1735-25G PC1735-100G
APOSPC1735-5GEA 21.9 EUR
APOSPC1735-5G APOSPC1735-25G APOSPC1735-100G
5-Fluoroxindol 98%
5-Fluoroxindol
Formel: C₈H₆FNO
Molecular Weight: 151,14 g/mol
Schmelzpunkt: 142…147 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179598
CAS-Nummer: 56341-41-4
EINECS: 446-540-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR