Methyl-2-brom-4-fluorbenzoat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-bromo-4-fluorobenzoate

PC1747-25G PC1747-5G PC1747-100G
APOSPC1747-25GEA 115 EUR
APOSPC1747-25G APOSPC1747-5G APOSPC1747-100G
Methyl-2-brom-4-fluorbenzoat 97%
Methyl-2-brom-4-fluorbenzoat
Formel: C₈H₆BrFO₂
Molecular Weight: 233,04 g/mol
Siedepunkt: 75…78 °C/1mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD06204026
CAS-Nummer: 653-92-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR