1-Chloro-1,1-difluoro-2,4-pentanedione 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Chloro-1,1-difluoroacetylacetone
PC1753G-25G
PC1753G-5G
APOSPC1753G-25GEA
343
EUR
APOSPC1753G-25G
APOSPC1753G-5G
1-Chloro-1,1-difluoro-2,4-pentanedione 97%
1-Chloro-1,1-difluoro-2,4-pentanedione
Formel:
C₅H₅ClF₂O₂ Molecular Weight: 170.54 g/mol Siedepunkt: 141 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041520 CAS-Nummer: 2375-76-0 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...