Kalium(4-(trifluormethyl)phenyl)trifluoroborat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Potassium 4-(trifluoromethyl)phenyltrifluoroborate, (T-4)-Potassium trifluoro(4-(trifluoromethyl)phenyl)borate

PC1754-5G PC1754-25G
APOSPC1754-5GEA 121 EUR
APOSPC1754-5G APOSPC1754-25G
Kalium(4-(trifluormethyl)phenyl)trifluoroborat 98%
Potassium-4-(trifluoromethyl)phenyltrifluoroborate
Formel: C₇H₄BF₆K
Molecular Weight: 252.01 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039955
CAS-Nummer: 166328-08-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR