3-(Ethoxycarbonyl)-5-fluorphenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-(Ethoxycarbonyl)-5-fluorophenyl)boronic acid, 3-Fluoro-5-(ethoxycarbonyl)benzeneboronic acid

PC1778-1G PC1778-5G
APOSPC1778-1GEA 249 EUR
APOSPC1778-1G APOSPC1778-5G
3-(Ethoxycarbonyl)-5-fluorphenylboronsäure 97%
3-(Ethoxycarbonyl)-5-fluorphenylboronsäure
Formel: C₉H₁₀BFO₄
Molecular Weight: 211,9800 g/mol
Schmelzpunkt: 148150 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07363792
CAS-Nummer: 871329-85-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR