3-Benzyloxy-5-fluorphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Benzyloxy-5-fluorobenzeneboronic acid
PC1786-1G
PC1786-5G
PC1786-10G
APOSPC1786-1GEA
21.9
EUR
APOSPC1786-1G
APOSPC1786-5G
APOSPC1786-10G
3-Benzyloxy-5-fluorphenylboronsäure 98%
3-Benzyloxy-5-fluorphenylboronsäure
Formel:
C₁₃H₁₂BFO₃ Molecular Weight: 246.04 g/mol Schmelzpunkt: 156…162 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07363782 CAS-Nummer: 850589-56-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...