3-Fluor-4-methoxybenzylchlorid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Fluoro-4-methoxybenzylchloride
PC1802-1G
PC1802-25G
PC1802-5G
APOSPC1802-1GEA
21.9
EUR
APOSPC1802-1G
APOSPC1802-25G
APOSPC1802-5G
3-Fluor-4-methoxybenzylchlorid
3-Fluor-4-methoxybenzylchlorid
Formel:
C₈H₈ClFO Molecular Weight: 174,60 g/mol Schmelzpunkt: 34…39 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973773 CAS-Nummer: 351-52-0 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...