3-Cyan-6-fluoranilin
Lieferant: Apollo Scientific
|
Achtung
|
PC1805-25G
APOSPC1805-25GEA
88.6
EUR
APOSPC1805-25G
3-Cyan-6-fluoranilin
3-Cyan-6-fluoranilin
Formel:
C₇H₅FN₂ Molecular Weight: 136,13 g/mol Schmelzpunkt: 90…92 °C Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07787428 CAS-Nummer: 859855-53-1 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...