4-Chlor-3-fluorbenzaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC1828M-25G
PC1828M-5G
PC1828M-1G
APOSPC1828M-25GEA
79.9
EUR
APOSPC1828M-25G
APOSPC1828M-5G
APOSPC1828M-1G
4-Chlor-3-fluorbenzaldehyd 98%
4-Chloro-3-fluorobenzaldehyde
Formel:
C₇H₄ClFO Molecular Weight: 158.56 g/mol Schmelzpunkt: 46…49 °C Flammpunkt: 96.7 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143288 CAS-Nummer: 5527-95-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...