4-Chlor-2-fluorbenzaldehyd 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1831-25G
PC1831-100G
PC1831-500G
APOSPC1831-25GEA
26.1
EUR
APOSPC1831-25G
APOSPC1831-100G
APOSPC1831-500G
4-Chlor-2-fluorbenzaldehyd 97%
4-Chloro-2-fluorobenzaldehyde
Formel:
C₇H₄ClFO Schmelzpunkt: 58…61 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143282 CAS-Nummer: 61072-56-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...