2-Chlor-4-fluorbenzamid 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1832-1G
PC1832-5G
PC1832-10G
PC1832-25G
APOSPC1832-1GEA
26.1
EUR
APOSPC1832-1G
APOSPC1832-5G
APOSPC1832-10G
APOSPC1832-25G
2-Chlor-4-fluorbenzamid 97%
2-Chlor-4-fluorbenzamid
Formel:
C₇H₅ClFNO Schmelzpunkt: 147…149 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042531 CAS-Nummer: 88578-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...