2-Chlor-6-fluorbenzamid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chlor-6-fluorbenzoesäureamid

PC1834-10G PC1834-1G PC1834-5G
APOSPC1834-10GEA 51 EUR
APOSPC1834-10G APOSPC1834-1G APOSPC1834-5G
2-Chlor-6-fluorbenzamid 97%
2-Chlor-6-fluorbenzamid
Formel: C₇H₅ClFNO
Molecular Weight: 173.57 g/mol
Schmelzpunkt: 139…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00055436
CAS-Nummer: 66073-54-9
EINECS: 266-123-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR