3-Chlor-4-fluorbenzamid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chlor-4-fluorbenzoesäureamid

PC1837B-10G PC1837B-1G PC1837B-5G
APOSPC1837B-10GEA 52.2 EUR
APOSPC1837B-10G APOSPC1837B-1G APOSPC1837B-5G
3-Chlor-4-fluorbenzamid 97%
3-Chlor-4-fluorbenzamid
Formel: C₇H₅ClFNO
Molecular Weight: 173.57 g/mol
Schmelzpunkt: 145…147 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00153092
CAS-Nummer: 701-43-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR