4-Fluor-o-toluonitril
Lieferant: Apollo Scientific
|
Achtung
|
PC1841-25G
PC1841-5G
PC1841-100G
APOSPC1841-25GEA
24.8
EUR
APOSPC1841-25G
APOSPC1841-5G
APOSPC1841-100G
4-Fluor-o-toluonitril
4-Fluor-o-toluonitril
Formel:
C₈H₆FN Molecular Weight: 135,14 g/mol Schmelzpunkt: 70…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095106 CAS-Nummer: 147754-12-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...