2',3'-Dichlor-6'-fluoracetophenon 98%

Lieferant: Apollo Scientific

PC1848-10G PC1848-5G PC1848-1G
APOSPC1848-10GEA 186 EUR
APOSPC1848-10G APOSPC1848-5G APOSPC1848-1G
2',3'-Dichlor-6'-fluoracetophenon 98%
2',3'-Dichlor-6'-fluoracetophenon
Formel: C₈H₅Cl₂FO
Molecular Weight: 207,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06798121
CAS-Nummer: 870704-16-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR