6-Chlor-5-fluorbenzimidazol 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloro-5-fluoro-1H-benzo[d]imidazole

PC1860F-1G PC1860F-250MG
APOSPC1860F-1GEA 132 EUR
APOSPC1860F-1G APOSPC1860F-250MG
6-Chlor-5-fluorbenzimidazol 95+%
6-Chloro-5-fluorobenzimidazole
Formel: C₇H₄ClFN₂
Molecular Weight: 170,56 g/mol
Schmelzpunkt: 148…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00156132
CAS-Nummer: 175135-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR