3-Chlor-4-fluorbenzonitril 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Chlor-3-cyan-6-fluorbenzene

PC1863D-25G PC1863D-100G
APOSPC1863D-25GEA 21.9 EUR
APOSPC1863D-25G APOSPC1863D-100G
3-Chlor-4-fluorbenzonitril 99%
3-Chlor-4-fluorbenzonitril
Formel: C₇H₃ClFN
Molecular Weight: 155,56 g/mol
Schmelzpunkt: 67…69 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015431
CAS-Nummer: 117482-84-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR