4-Chloro-2-fluorobenzylamine hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Chloro-2-fluorophenyl)methanamine hydrochloride

PC1864FD-1G PC1864FD-25G PC1864FD-5G
APOSPC1864FD-1GEA 21.9 EUR
APOSPC1864FD-1G APOSPC1864FD-25G APOSPC1864FD-5G
4-Chloro-2-fluorobenzylamine hydrochloride 97%
4-Chloro-2-fluorobenzylamine hydrochloride
Formel: C₇H₇ClFN·HCl
Molecular Weight: 196,05 g/mol
Schmelzpunkt: 243…245 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143286
CAS-Nummer: 202982-63-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR