4-Chlor-2-fluorzimtsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(4-Chlor-2-fluorphenyl)propensäure, 4-Chlor-2-fluorcinnamsäure

PC1864R-25G PC1864R-5G PC1864R-1G
APOSPC1864R-25GEA 74.1 EUR
APOSPC1864R-25G APOSPC1864R-5G APOSPC1864R-1G
4-Chlor-2-fluorzimtsäure 98%
4-Chlor-2-fluorzimtsäure
Formel: C₉H₆ClFO₂
Molecular Weight: 200.6 g/mol
Schmelzpunkt: 215…218 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143287
CAS-Nummer: 202982-65-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR