5-Chlor-4-fluor-2-nitroanilin 97+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Amino-3-chlor-4-fluor-6-nitrobenzene
PC1868R-1G
PC1868R-25G
PC1868R-5G
APOSPC1868R-1GEA
58.2
EUR
APOSPC1868R-1G
APOSPC1868R-25G
APOSPC1868R-5G
5-Chlor-4-fluor-2-nitroanilin 97+%
5-Chlor-4-fluor-2-nitroanilin
Formel:
C₆H₄ClFN₂O₂ Molecular Weight: 190,56 g/mol Schmelzpunkt: 142…147 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00052698 CAS-Nummer: 104222-34-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...