4-Nitro-α,α,α-trifluor-o-toluylsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Nitro-2-(trifluormethyl)benzoesäure
PC18759-25G
PC18759-5G
PC18759-10G
APOSPC18759-25GEA
53.8
EUR
APOSPC18759-25G
APOSPC18759-5G
APOSPC18759-10G
4-Nitro-α,α,α-trifluor-o-toluylsäure
4-Nitro-α,α,α-trifluor-o-toluylsäure
Formel:
C₈H₄F₃NO₄ Molecular Weight: 235,12 g/mol Schmelzpunkt: 135…142 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039246 CAS-Nummer: 320-37-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...