2-(3-Chlor-4-fluorphenyl)indol 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1884T-1G
PC1884T-5G
APOSPC1884T-1GEA
161
EUR
APOSPC1884T-1G
APOSPC1884T-5G
2-(3-Chlor-4-fluorphenyl)indol 97%
2-(3-Chloro-4-fluorophenyl)indole
Formel:
C₁₄H₉ClFN Schmelzpunkt: 170…173 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00068179 CAS-Nummer: 1868-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...