3-[2-(Trifluoromethyl)phenyl]propionic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-[2-(Trifluoromethyl)phenyl]propanoic acid
PC1935-25G
PC1935-5G
PC1935-10G
APOSPC1935-25GEA
81.2
EUR
APOSPC1935-25G
APOSPC1935-5G
APOSPC1935-10G
3-[2-(Trifluoromethyl)phenyl]propionic acid 97%
3-[2-(Trifluoromethyl)phenyl]propionic acid
Formel:
C₁₀H₉F₃O₂ Molecular Weight: 218.17 g/mol Schmelzpunkt: 84…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06823965 CAS-Nummer: 94022-99-8 EINECS: 301-594-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...