2-Fluor-4-methoxyphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-4-methoxybenzeneboronic acid

PC1942-1G PC1942-5G
APOSPC1942-1GEA 21.9 EUR
APOSPC1942-1G APOSPC1942-5G
2-Fluor-4-methoxyphenylboronsäure
2-Fluor-4-methoxyphenylboronsäure
Formel: C₇H₈BFO₃
Molecular Weight: 169,95 g/mol
Schmelzpunkt: 117,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039323
CAS-Nummer: 162101-31-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR