Methyl 3-Amino-2-fluorobenzoate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 3-amino-2-fluorobenzoate, 3-Amino-2-fluorobenzoic acid methyl ester
PC19685-250MG
PC19685-1G
APOSPC19685-250MGEA
33.5
EUR
APOSPC19685-250MG
APOSPC19685-1G
Methyl 3-Amino-2-fluorobenzoate
Methyl 3-Amino-2-fluorobenzoate
Formel:
C₈H₈FNO₂ Molecular Weight: 169,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12923265 CAS-Nummer: 1195768-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...