6-(Trifluormethyl)uracil 98%
Lieferant: Apollo Scientific
Synonyme:
4-(Trifluormethyl)uracil, 2,4-Dihydroxy-6-(trifluormethyl)pyrimidin, 2,6-Dihydroxy-4-(trifluormethyl)pyrimidin
PC1992-1G
PC1992-5G
PC1992-25G
APOSPC1992-1GEA
102
EUR
APOSPC1992-1G
APOSPC1992-5G
APOSPC1992-25G
6-(Trifluormethyl)uracil 98%
6-(Trifluormethyl)uracil
Formel:
C₅H₃F₃N₂O₂ Molecular Weight: 180.09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01011762 CAS-Nummer: 672-45-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...