2-Amino-5-fluorbenzamid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Fluoroanthranilamide
PC1994-1G
PC1994-250MG
PC1994-5G
APOSPC1994-1GEA
87.3
EUR
APOSPC1994-1G
APOSPC1994-250MG
APOSPC1994-5G
2-Amino-5-fluorbenzamid 97%
2-Amino-5-fluorbenzamid
Formel:
C₇H₇FN₂O Molecular Weight: 154,14 g/mol Schmelzpunkt: 146…148 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03428522 CAS-Nummer: 63069-49-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...