Ethyl-N-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]carbamothioyl}carbamate

Lieferant: Apollo Scientific

PC200004-1G PC200004-500MG
APOSPC200004-1GEA 111 EUR
APOSPC200004-1G APOSPC200004-500MG
Ethyl-N-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]carbamothioyl}carbamate
Ethyl-N-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]carbamothioyl}carbamate
Formel: C₁₀H₉ClF₃N₃O₂S
Molecular Weight: 327,71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22383949
CAS-Nummer: 1206641-15-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR