N-(2-Amino-4-fluoro-6-methylphenyl)acetamide

Lieferant: Apollo Scientific

PC200005-1G PC200005-500MG
APOSPC200005-1GEA 311 EUR
APOSPC200005-1G APOSPC200005-500MG
N-(2-Amino-4-fluoro-6-methylphenyl)acetamide
N-(2-Amino-4-fluoro-6-methylphenyl)acetamide
Formel: C₉H₁₁FN₂O
Schmelzpunkt: 144…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD22573532
CAS-Nummer: 1407516-46-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR