Ethyl-2,6-dichloro-5-fluoronicotinate 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2,6-dichloro-5-fluoropyridine-3-carboxylate
PC200006-1G
PC200006-5G
PC200006-10G
APOSPC200006-1GEA
65.4
EUR
APOSPC200006-1G
APOSPC200006-5G
APOSPC200006-10G
Ethyl-2,6-dichloro-5-fluoronicotinate 97%
Ethyl-2,6-dichloro-5-fluoronicotinate
Formel:
C₈H₆Cl₂FNO₂ Molecular Weight: 238.05 g/mol Siedepunkt: 95…97 °C/2…3mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD09757511 CAS-Nummer: 82671-03-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...