1-Bromo-2-(chloromethyl)-4-fluorobenzene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-5-fluorobenzylchloride, 2-Bromo-5-fluorobenzyl chloride
PC200007-1G
PC200007-500MG
PC200007-5G
APOSPC200007-1GEA
168
EUR
APOSPC200007-1G
APOSPC200007-500MG
APOSPC200007-5G
1-Bromo-2-(chloromethyl)-4-fluorobenzene
1-Bromo-2-(chloromethyl)-4-fluorobenzene
Formel:
C₇H₅BrClF Molecular Weight: 223,48 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09263441 CAS-Nummer: 857276-61-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...