(4-Chlorothiophen-2-yl)(4-fluorophenyl)methanone

Lieferant: Apollo Scientific

PC200010-1G PC200010-500MG
APOSPC200010-1GEA 336 EUR
APOSPC200010-1G APOSPC200010-500MG
(4-Chlorothiophen-2-yl)(4-fluorophenyl)methanone
(4-Chlorothiophen-2-yl)(4-fluorophenyl)methanone
Formel: C₁₁H₆ClFOS
Molecular Weight: 240.69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22573546
CAS-Nummer: 207852-56-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR