Methyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-2-cyanoacetate

Lieferant: Apollo Scientific

PC200012-1G PC200012-500MG PC200012-5G
APOSPC200012-1GEA 102 EUR
APOSPC200012-1G APOSPC200012-500MG APOSPC200012-5G
Methyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-2-cyanoacetate
Methyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-2-cyanoacetate
Formel: C₁₀H₆ClF₃N₂O₂
Molecular Weight: 278,62 g/mol
Schmelzpunkt: 206…208 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16622814
CAS-Nummer: 658066-43-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR