(4-Fluoro-3-iodophenyl)methanol 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluoro-3-iodobenzyl alcohol

PC200014-1G PC200014-250MG PC200014-5G
APOSPC200014-1GEA 227 EUR
APOSPC200014-1G APOSPC200014-250MG APOSPC200014-5G
(4-Fluoro-3-iodophenyl)methanol 96%
(4-Fluoro-3-iodophenyl)methanol
Formel: C₇H₆FIO
Molecular Weight: 252.03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18396949
CAS-Nummer: 227609-87-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR