Methyl-7-(trifluoromethyl)-1-benzofuran-2-carboxylate

Lieferant: Apollo Scientific

PC200024-1G PC200024-500MG
APOSPC200024-1GEA 311 EUR
APOSPC200024-1G APOSPC200024-500MG
Methyl-7-(trifluoromethyl)-1-benzofuran-2-carboxylate
Methyl-7-(trifluoromethyl)-1-benzofuran-2-carboxylate
Formel: C₁₁H₇F₃O₃
Molecular Weight: 244.17 g/mol
Schmelzpunkt: 51…53 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD22573541
CAS-Nummer: 1407521-92-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR