2,2,2-Trifluoro-3'-nitroacetophenone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3'-Nitro-2,2,2-trifluoroacetophenone, 2,2,2-Trifluoro-1-(3-nitrophenyl)ethanone
PC200041-1G
PC200041-25G
APOSPC200041-1GEA
62.5
EUR
APOSPC200041-1G
APOSPC200041-25G
2,2,2-Trifluoro-3'-nitroacetophenone
2,2,2-Trifluoro-3'-nitroacetophenone
Formel:
C₈H₄F₃NO₃ Molecular Weight: 219,12 g/mol Schmelzpunkt: 55…57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00034541 CAS-Nummer: 657-15-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...