8-Chloro-6-(trifluoromethyl)-4(1H)-quinolinone 95+%

Lieferant: Apollo Scientific
Achtung

PC200056-1G PC200056-250MG
APOSPC200056-1GEA 499 EUR
APOSPC200056-1G APOSPC200056-250MG
8-Chloro-6-(trifluoromethyl)-4(1H)-quinolinone 95+%
8-Chloro-6-(trifluoromethyl)-4(1H)-quinolinone
Formel: C₁₀H₅ClF₃NO
Molecular Weight: 247,61 g/mol
Schmelzpunkt: 285…286 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12827776
CAS-Nummer: 1065092-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR