3-Amino-4-fluor-1H-indazol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluoro-1H-indazol-3-amine

PC200069-5G
APOSPC200069-5GEA 144 EUR
APOSPC200069-5G
3-Amino-4-fluor-1H-indazol
3-Amino-4-fluoro-1H-indazole
Formel: C₇H₆FN₃
Molecular Weight: 151,14 g/mol
Schmelzpunkt: 172…174 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00119934
CAS-Nummer: 404827-78-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR